Learn More
DWK LIFE SCIENCES™ WHEATON™ E-Z Seals With PTFE/Grey Butyl Liner
SDP
Catalog No. 06451117 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Natural
Catalog No. Color
06-451-117 Natural
1 options
Catalog No. 06-451-117 Supplier DWK LIFE SCIENCES™ Supplier No. 22423501
Add to Cart
Add to Cart
WHEATON™ E-Z Seals With PTFE/Grey Butyl Liner provide a reliable crimp seal solution for 11mm vials. Featuring a PTFE/Grey Butyl liner, these aluminum seals help maintain sample integrity and are available in multiple colors for easy identification.
  • 11mm aluminum crimp seals designed for use with standard 11mm vials
  • PTFE/Grey Butyl liner construction helps protect sample integrity
  • Available in multiple seal colors for convenient vial identification
  • Sold in shelf packs of 100; 1000 per case
  • Wheaton brand quality from DWK Life Sciences
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Butyl
Color Natural
Diameter (Metric) Outer 11 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum PTFE/Grey Butyl
Product Line E-Z Seal
Quantity 1000/Cs.
Closure Size 11 mm
Autoclavable Not Autoclavable
Product Type Open Top Cap