Learn More
DWK Life Sciences Wheaton™ Extended Controlled 20mm LDPE Dropper Tips
Shop All DWK Life Sciences Products

Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
- Case of 100
Specifications
Specifications
| Color | Green |
| Material | LDPE |
| No. per Case | 100 Cs. |
| Drop Size | 41 μL |
| For Use With (Equipment) | Plastic dropping bottle |
| Certifications/Compliance | ISO 9001 |
| Product Type | Dropper Tip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.