Learn More
DWK Life Sciences WHEATON™ Glass Sample Vials With Metal Foil Caps
Shop All DWK Life Sciences Products

Description
- Made from borosilicate glass conforming to USP Type I and ASTM E 438 Type I, Class A requirements
- Metal foil caps attached to vials for secure, ready-to-use sample containment
- Suitable for buffers, reagents, and liquid, solid, and semi-solid content
- Background counts are consistent and low; ultraviolet transmission is high
- Ideal for sample storage and laboratory analysis applications
Specifications
Specifications
| Material (Liner) | Metal Foil |
| Color | Clear |
| Diameter (Metric) Outer | 12 mm |
| Material | Borosilicate Glass (Low Extractable) |
| Pre-slit | No |
| Sterility | Non-sterile |
| Volume (Metric) | 2 mL |
| For Use With (Application) | Used for storing and transporting vials |
| Thread Style | Screw |
| Volume (English) | 0.5 dr. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.