Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Glass Serum Bottles
SDP

Catalog No. 0645142 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
06-451-42 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-451-42 Supplier DWK Life Sciences Supplier No. 223950

missing translation for 'validMainframeMsg'

Graduated Serum Bottles are manufactured from WHEATON from 400 borosilicate glass that conforms to USP Type I requirements

  • Raised graduations indicate approximate capacities in milliliters
  • Accepts standard stoppers and seals
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Color Clear
Diameter (English) 2.36 in.
Diameter (Metric) Outer Neck 30mm
Diameter (Metric) Inner Neck 15mm
Material Borosilicate Glass
Height (Metric) 158 mm
Height (English) 6.22 in.
Diameter (Metric) 60 mm
Product Type Serum Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.