Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Replacement Screw Cap for Coplin Staining Jar 900520/900570, 58-400 Finish
GSA_VASDP

Catalog No. 02912618 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
5 to 10 Slide Staining Dish
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
02-912-618 5 to 10 Slide Staining Dish
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-912-618 Supplier DWK Life Sciences Supplier No. 900522
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Replacement Screw Cap for Coplin Staining Jar 900520/900570, 58-400 Finish is a linerless white polypropylene cap designed to reduce solvent evaporation and spills. Compatible with WHEATON™ Coplin staining jars. Cap is not intended for use with slides in jar.

  • Replacement screw cap (58-400 finish) for Wheaton Coplin Staining Jars 900520 and 900570
  • Made of linerless white polypropylene to reduce solvent evaporation and spills during storage
  • Cap is not intended for use with slides in jar
  • Sold as 6 per case
  • Supplied by DWK Life Sciences under the Wheaton brand

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Type Screw
Autoclavable Not Autoclavable
Color White
Material Polypropylene
Type Screw Cap
For Use With (Equipment) 5 to 10 Slide Staining Dish
Height (Metric) 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.