Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Green Starline Polypropylene Caps for Redi-Pak™ Narrow-Mouth Containers
SDP

Catalog No. 0987710E Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
20-410
28-410
No. per Case:
48 Cs.
72 Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
09-877-10E 13-425 72 Cs.
09-877-11E 20-410 72 Cs.
09-877-13E 28-410 48 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-877-10E Supplier DWK Life Sciences Supplier No. 23950106
Add to Cart
Add to Cart

Caps that are the perfect fit for WHEATON Leak Resistant Narrow Mouth Bottles

  • Leak resistant
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.