Learn More
DWK Life Sciences Wheaton™ Hose Connector
SDP
Catalog No. 1420993 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
13-425 GPI
Catalog No. Thread Size
14-209-93 13-425 GPI
1 options
Catalog No. 14-209-93 Supplier DWK Life Sciences Supplier No. 962515
Add to Cart
Add to Cart
Grease-free fitting for condensers and connection to a vacuum
  • For continuous extraction of analytes
  • All wetted parts are TFE
  • Unscrew to disconnect, leaving hose attached
  • O.D of smallest serration:0.375 in. (9.5mm)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material PTFE
For Use With (Application) For continuous extraction of analytes and micro soxhlet extraction apparatus
Thread Size 13-425 GPI
Product Type Hose Connector