Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Evaporative Flask
SDP

Catalog No. 02912565 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-912-565 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-912-565 Supplier DWK Life Sciences Supplier No. 297825
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For concentration of trace amounts of analyte in organic solvent

  • 24/40 top and 19/22 bottom clear seal joints
TRUSTED_SUSTAINABILITY

Specifications

Product Type Evaporative Flask
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Includes 24/40 top and 19/22 bottom clearseal joints
For Use With (Application) Concentration of trace amounts of analyte in organic solvent
Certifications/Compliance ASTM E 438
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.