Learn More
DWK Life Sciences Wheaton™ Kuderna-Danish Evaporative Concentrator With Clear-Seal™ Joints
SDP
Catalog No. 02912563 Shop All DWK Life Sciences Products
Change view
Click to view available options
Certifications/Compliance:
ASTM E 438
Catalog No. Certifications/Compliance
02-912-563 ASTM E 438
1 options
Catalog No. 02-912-563 Supplier DWK Life Sciences Supplier No. 297883
Add to Cart
Add to Cart
For concentration of trace amounts of analyte in organic solvent
  • Clear seal joint
TRUSTED_SUSTAINABILITY
Specifications
Product Type Kuderna-Danish Evaporative Concentrator
Includes Clear seal joint
For Use With (Application) Concentration of trace amounts of analyte in organic solvent
Certifications/Compliance ASTM E 438