Learn More
DWK Life Sciences WHEATON™ LDPE Dropping Bottles with Foamed Polyethylene Lined Caps
Shop All DWK Life Sciences Products

Description
- Low-density polyethylene (LDPE) construction with translucent color for easy content visibility
- LDPE material is very flexible with high impact strength
- Narrow mouth design ideal for dispensing liquids with control
- Pre-attached white polypropylene screw cap with foamed polyethylene liner for a secure, leak-resistant seal
- Good chemical resistance suitable for mild and strong buffers
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Color | White |
| Diameter (English) | 1.5 in. |
| Includes | Foam Polyethylene lined cap |
| Material | LDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Narrow |
| Height (Metric) | 88 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.