Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Low-Density Extended Controlled Dropper Tips 13 mm
SDP

Catalog No. 0344055 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material No. per Case
03-440-55 Orange LDPE 1000 Cs.
03-440-54 Blue LDPE 1000 Cs.
06-450-122 Blue LDPE 100 Cs.
03-440-53 Gray Polyethylene 1000 Cs.
03-440-52 Green LDPE 1000 Cs.
03-440-58 Natural LDPE 1000 Cs.
06-450-121 Orange LDPE 100 Cs.
03-440-57 Pink LDPE 1000 Cs.
06-450-119 Pink LDPE 100 Cs.
03-440-51 Purple Polyethylene 1000 Cs.
06-450-125 Purple LDPE 100 Cs.
03-440-56 Red LDPE 1000 Cs.
06-450-120 Red LDPE 100 Cs.
03-440-48 White Polyethylene 1000 Cs.
03-440-50 Yellow LDPE 1000 Cs.
06-450-126 Yellow LDPE 100 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 03-440-55 Supplier DWK Life Sciences Supplier No. W242413A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Low-Density Extended Controlled Dropper Tips (13 mm) are made of LDPE and dispense one drop at a time with an average drop size of 44μL ±6μL. Available in multiple colors, these 13mm tips offer reliable, repeatable dispensing for biotech, diagnostic, and pharmaceutical use.

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Made of Low Density Polyethylene (LDPE) for excellent chemical resistance
  • 13 mm tip size fits standard dropper bottle closures
  • Available in multiple colors for easy identification
  • Ideal for biotech, diagnostic, and pharmaceutical applications
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material LDPE
No. per Case 1000 Cs.
Quantity 1,000/Cs.
Sterility Non-sterile
Drop Size 41 μL
For Use With (Equipment) Plastic dropping bottle
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Dropper Tip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.