Learn More
Electron Microscopy Sciences Wheaton M-T/40 Vial File
SDP
Catalog No. 501898993
Encompass_Preferred
This filing system offers a clean, compact, and convenient alternative for the storage of biological samples. A clear polystyrene case, with hinged lid, alpha-numerical index, and individual partitions. Foam inserts are provided as well and they offer extra protection for the vials. Stores up to 40 vials of 16mm diameter x 50mm height.
Catalog No. 50-189-8993 Supplier Electron Microscopy Sciences Supplier No. 6099046
This filing system offers a clean, compact, and convenient alternative for the storage of biological samples. A clear polystyrene case, with hinged lid, alpha-numerical index, and individual partitions. Foam inserts are provided as well and they offer extra protection for the vials. Stores up to 40 vials of 16mm diameter x 50mm height.