Learn More
Electron Microscopy Sciences Wheaton M-T-60 Vial File, 6¾" (L) x 3¾" (W) x 2½" (H) (190 x 95 x 62 mm)
SDP
Catalog No. 501898994
Encompass_Preferred
This filing system offers a clean, compact, and convenient alternative for the storage of biological samples. A clear polystyrene case, with hinged lid, alpha-numerical index, and individual partitions. Foam inserts are provided as well and they offer extra protection for the vials. Stores up to 60 vials of 13mm diameter x 50mm height.
Catalog No. 50-189-8994 Supplier Electron Microscopy Sciences Supplier No. 6099060
This filing system offers a clean, compact, and convenient alternative for the storage of biological samples. A clear polystyrene case, with hinged lid, alpha-numerical index, and individual partitions. Foam inserts are provided as well and they offer extra protection for the vials. Stores up to 60 vials of 13mm diameter x 50mm height.