Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton media bottles glas
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z36483524EA
Encompass
Low-alkali borosilicate glass bottles especially suitable for storing culture media. No-drip lip for clean pouring sloping shoulder for easier cleaning. Graduations show approximate volume. Large writing area.
Catalog No. 50-290-5724