Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton media bottles glas
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z36485148EA
Low-alkali borosilicate glass bottles especially suitable for storing culture media. No-drip lip for clean pouring sloping shoulder for easier cleaning. Graduations show approximate volume. Large writing area.