Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton media bottles glas
Catalog No. 502905865
Encompass
Low-alkali borosilicate glass bottles especially suitable for storing culture media. No-drip lip for clean pouring sloping shoulder for easier cleaning. Graduations show approximate volume. Large writing area.
Catalog No. 50-290-5865 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z36481948EA
Low-alkali borosilicate glass bottles especially suitable for storing culture media. No-drip lip for clean pouring sloping shoulder for easier cleaning. Graduations show approximate volume. Large writing area.