Learn More
Socorex™ WHEATON™ Micro Kit and Accessories
SDP
Catalog No. 1532014B Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Claisen Adapter
Connecting Adapter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-320-14B Connecting Adapter
11-302-32C Claisen Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-320-14B Supplier Socorex™ Supplier No. 123753
Add to Cart
Add to Cart
WHEATON™ Micro Kit and Accessories features Clear-Seal™ grease-free joints with smooth, unground surfaces that seal without grease, reducing seizing and contamination. Ideal for a wide range of organic chemistry experiments.
  • Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately
  • WHEATON standard taper Clear-Seal™ grease-free joints provide smooth, unground surfaces, which seal without grease and reduce seizing and contamination from grease
  • Clear-Seal™ joints can replace ground units in any application and provide other important advantages over ground joints
  • Superior in vacuum and radioactive applications
  • Smooth surfaces resist contamination
  • Cleaning is simple and thorough
  • Interchangeable with ground joints
  • All chemical changes and thermometer scales remain visible through the joints
  • Clear-Seal joints have never been weakened by grinding and are formed from special wall tubing that is heavier than standard wall for extra strength
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
  • Clear-Seal joints are produced to American Standard Taper dimensions and can be used interchangeably with any standard taper ground joints
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Wheaton micro kit
Includes 19/22 Clear-Seal™ joints
Material Borosilicate Glass (33 Low Extractable)
Product Type Connecting Adapter
Standard Taper Joint No. 19/22