Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton narrow mouth bottl
Catalog No. 502901869
Encompass
Convenience packed with closures attached. Bottles made of Wheaton 900 amber glass or 800 clear soda lime flint glass.Choice of cap stylesWhite PP cap with PTfE or polyvinyl linerBlack plastic cap with Poly-Seal(R) PE cone liner
Catalog No. 50-290-1869 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2500231PAK
Convenience packed with closures attached. Bottles made of Wheaton 900 amber glass or 800 clear soda lime flint glass.Choice of cap stylesWhite PP cap with PTfE or polyvinyl linerBlack plastic cap with Poly-Seal(R) PE cone liner