Learn More
DWK Life Sciences WHEATON™ One-Piece Scrubber Absorber Tube
SDP
Catalog No. 0140511 Shop All DWK Life Sciences Products
Change view
Click to view available options
Quantity:
1/Cs.
Catalog No. Quantity
01-405-11 1/Cs.
1 options
Catalog No. 01-405-11 Supplier DWK Life Sciences Supplier No. 180024
Add to Cart
Add to Cart
The WHEATON™ One-Piece Scrubber Absorber Tube is a modified version of catalog number 180020, engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method. Meets ASTM, USP, and EPA specifications.
  • Modified one-piece design eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes
  • Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
  • Meets ASTM, USP, and EPA specifications
  • Features a standard taper 24/40 Clear-Seal™ joint for a secure, freestanding system
  • Manufactured from WHEATON 33 low extractable borosilicate glass conforming to USP Type I and ASTM E 438 Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Arsine Generator
Standard Taper Joint 24/40
Material Borosilicate Glass
For Use With (Application) For arsenic analysis
Quantity 1/Cs.