Learn More
DWK Life Sciences WHEATON™ One–Sep Polypropylene Septas
SDP
Catalog No. 0334093 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
03-340-93 Black
1 options
Catalog No. 03-340-93 Supplier DWK Life Sciences Supplier No. 242247
Add to Cart
Add to Cart
WHEATON™ One–Sep Polypropylene Septas are designed to fit screw caps with 13–425 GPI thread. Made from polypropylene, these black septas provide a reliable seal for laboratory vials and sample containers.
  • Fits screw caps with 13–425 GPI thread
  • Cap color: Black
  • Cap material: Polypropylene
  • Liner material: Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polypropylene
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable