Learn More
DWK Life Sciences Wheaton™ Overhead Stirrer Accessories - Extension Sleeve
SDP
Catalog No. 02911543 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Extension Sleeve
Catalog No. Type
02-911-543 Extension Sleeve
1 options
Catalog No. 02-911-543 Supplier DWK Life Sciences Supplier No. 905475
Add to Cart
Add to Cart
Includes
Aluminum rod and clamp for unit mounting
Extension sleeves, rods and Swivel blade paddle for Overhead Stirrer
  • Dual purpose unit for use with tissue grinders or impellers
  • Adjustable speed ranging from 300 to 5000 rpm under load
  • Adjustable connector accepts stainless steel shafts up to 0.3125 in. (7.9 mm) in diameter
Order Info
Electrical requirements: 120 VAC, 50/60 Hz, 50 watts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Extension sleeve for lengthening paddles
Frequency 50/60 Hz
Type Extension Sleeve
Wattage 50 w