Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Phenolic Screw Caps for Media Bottles
SDP

Catalog No. 22295763 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
40 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
22-295763 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-295763 Supplier DWK Life Sciences Supplier No. 240481
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Phenolic Screw Caps for Media Bottles feature a PTFE faced (14B) styrene-butadiene rubber liner that provides a totally inert inner seal and surface facing the sample or product. Autoclavable for repeated sterilization cycles.

  • Designed for the ultimate in product safety
  • Black phenolic screw cap construction
  • PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product
  • Autoclavable, withstands repeated sterilization cycles
  • Suitable for use with media bottles
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Sterility Non-sterile
Thread Size (Metric) 40 mm
Liner Material PTFE/Rubber
Neck Finish 38-430
For Use With (Equipment) Media bottle
Autoclavable Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Bottle Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.