Learn More
DWK Life Sciences WHEATON™ Dosys 172 Pistol Grip Basic Syringe
SDP
Catalog No. 13687450 Shop All DWK Life Sciences Products
Change view
Click to view available options
Needle Gauge:
14 G
Catalog No. Needle Gauge
13-687-450 14 G
1 options
Catalog No. 13-687-450 Supplier DWK Life Sciences Supplier No. W853006
Add to Cart
Add to Cart
The WHEATON™ Dosys 172 Pistol Grip Basic Syringe is a self-refilling automatic syringe with an integrated valve system and Luer Lock nozzle, delivering precise and repeatable dispensing across multiple volume ranges.
  • Self-refilling automatic syringe for precise, repeatable liquid dispensing
  • Integrated valve system ensures reliable and consistent performance
  • Luer Lock nozzle for secure connections
  • Pistol grip handle design for comfortable, controlled operation
  • Package includes syringe, one metre silicone feed tube, sinker, PVC protection sleeve, aspiration and vent cannulas, set of spares for valve, and operating instructions
  • Available in multiple volume ranges to suit a variety of laboratory applications
TRUSTED_SUSTAINABILITY
Specifications
Material Metal
Description Pistol Grip Syringe
Needle Gauge 14 G
Product Type Syringe
Volume (Metric) 10 mL
Connection Type Luer Lock
Graduated Yes
Sterility Non-sterile