Learn More
Description
- Ideal for mixing and storing culture media, distilled water, analytical standards and reagents
- Large opening with special lip and sloping shoulders for easy pouring and cleaning
- Fitted with polyethylene-lined screw caps for secure sealing
- Supplied with caps pre-attached and shrink-wrapped in convenient trays
- Sterile and ready for use
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Color | Clear |
| Diameter (English) | 2.08 in. |
| For Use With (Application) | For distilled Water, analytical standards and reagents when fitted with a Polyethylene lined screw cap |
| Material | PET |
| Quantity | 24/Cs. |
| Sterility | Sterile |
| Neck Finish | 33-430 |
| Closure Included | Yes |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

