Learn More
DWK Life Sciences Wheaton™ Plurascan Bar Code Reader
SDP
Catalog No. 03410049 Shop All DWK Life Sciences Products
Change view
Click to view available options
Certifications/Compliance:
CE
Catalog No. Certifications/Compliance
03-410-049 CE
1 options
Catalog No. 03-410-049 Supplier DWK Life Sciences Supplier No. W986010
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Capable of reading bar codes from wide range of manufacturers and label printers
Works with WHEATON KeepIT™ 81 and KeepIt 100™ Boxes and WHEATON CryoELITE™ Cryogenic Vials and Cryule™ Ampules
  • Integrated frost reduction system allows multiple racks to be read
  • Flexible software integration
Compliance
Conforms to CE Standards
Warranty and Services
One year
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Capable of reading 2D data matrix barcodes from wide range of manufacturers and label printers
Product Type Plurascan™ Barcode Reader
For Use With (Equipment) Wheaton™ KeepIT 81 and 100 boxes and Wheaton™ CryoElite™ cryogenic vials and cryule ampoules
Certifications/Compliance CE