Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Polypropylene Caps For WHEATON™ Dropping Bottles, Green
SDP

Catalog No. 06450184 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
06-450-184 13-425 100/Cs.
03-439-86 13-425 1000/Cs.
03-439-97 20-410 1000/Cs.
03-440-08 15-415 1000/Cs.
06-450-154 8-425 100/Cs.
06-450-164 15-415 100/Cs.
06-450-174 20-410 100/Cs.
03-440-18 8-425 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-450-184 Supplier DWK Life Sciences Supplier No. 242536
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Polypropylene Caps For WHEATON™ Dropping Bottles - Green are radiation grade polypropylene screw caps designed for use with WHEATON™ Dropping Bottles. Available in multiple sizes and pack quantities to suit laboratory needs.

  • Radiation grade polypropylene construction
  • Green colour for easy identification
  • Compatible with Wheaton Dropping Bottles
  • Available in sizes 8-425, 13-425, 15-415, and 20-410
  • Available in pack quantities of 100 or 1000 per case
TRUSTED_SUSTAINABILITY

Specifications

Color Purple
Material Polypropylene
No. per Case 100/Cs.
Sterility Non-sterile
Neck Finish 13-425
For Use With (Equipment) Plastic dropping bottle
Autoclavable Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Polypropylene Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.