Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles
SDP

Catalog No. 12141605 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
15-415
8-425
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
12-141-605 15-415
12-141-604 8-425
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-605 Supplier DWK Life Sciences Supplier No. 24251101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Screw cap made of radiation grade polypropylene.

  • Made from polypropylene offering good chemical resistance
  • Manufactured to stringent ISO and quality standards and Lot controlled
  • Choice of colors and sizes to suit most user needs
  • Packaged in case quantities of either 100 or 1000 pieces
  • Product is double bagged and twist tied to help reduce particulates
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
For Use With (Application) Sealing Dropper Tips on Dropping Bottles, Controlled Liquid Dispensing
Material Polypropylene
No. per Case 100
Sterility Non-sterile
Thread Size (Metric) 15-415
Fit for Purpose Certified Quality
For Use With (Equipment) Wheaton (15 mm) dropper tips and Wheaton dropping bottles
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Dropper Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.