Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Polypropylene Caps for WHEATON™ Dropping Bottles, Blue
SDP

Catalog No. 06450162 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
06-450-162 15-415 100/Cs.
03-439-99 20-410 1000/Cs.
06-450-152 8-425 100/Cs.
03-440-20 8-425 1000/Cs.
03-440-10 15-415 1000/Cs.
06-450-172 20-410 100/Cs.
03-439-88 13-425 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 06-450-162 Supplier DWK Life Sciences Supplier No. 242514
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Polypropylene Caps for WHEATON™ Dropping Bottles - Blue are screw caps made of radiation grade polypropylene. Available in multiple thread sizes and sold in cases of 100 or 1,000 to suit laboratory dispensing needs.

  • Screw cap made of radiation grade polypropylene
  • Blue color for easy identification
  • Compatible with Wheaton Dropping Bottles
  • Available in thread sizes 8-425, 13-425, 15-415, and 20-410
  • Packaged in cases of 100 or 1,000
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
No. per Case 100/Cs.
Sterility Non-sterile
Neck Finish 15-415
For Use With (Equipment) Plastic dropping bottle
Autoclavable Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Polypropylene Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.