Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Red
SDP

Catalog No. 06450150 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100 Cs.
1000 Cs.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Neck Finish No. per Case
06-450-150 Plastic dropping bottle 8-425 100 Cs.
03-439-90 Dropping bottle 13-425 1000 Cs.
03-440-01 Dropping bottle 20-410 1000 Cs.
03-440-12 Dropping bottle 15-415 1000 Cs.
03-440-22 Plastic dropping bottle 8-425 1000 Cs.
06-450-160 Plastic dropping bottle 15-415 100 Cs.
06-450-170 Plastic dropping bottle 20-410 100 Cs.
06-450-180 Plastic dropping bottle 13-425 100 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-450-150 Supplier DWK Life Sciences Supplier No. 242502
Add to Cart
Add to Cart

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Polypropylene
No. per Case 100 Cs.
Neck Finish 8-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.