Learn More
DWK Life Sciences WHEATON™ Polypropylene Caps for WHEATON™ Dropping Bottles, White
Shop All DWK Life Sciences Products
Description
- Radiation grade polypropylene construction
- Designed for use with Wheaton Dropping Bottles
- Available in multiple closure sizes: 8-425, 13-425, 15-415, and 20-410
- White color
- Available in cases of 100 or 1000
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| No. per Case | 100/Cs. |
| Sterility | Non-sterile |
| Neck Finish | 13-425 |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Polypropylene Cap |