Learn More
DWK Life Sciences WHEATON™ Polypropylene Clamps for Erlenmeyer Shake Flasks
SDP
Catalog No. 02924989 Shop All DWK Life Sciences Products
Change view
Click to view available options
Holds:
1000 mL Shaker Flask
125 mL Shaker Flask
2000 mL Shaker Flask
250 mL Shaker Flask
500 mL Shaker Flask
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
02-924-989 2000 mL Shaker Flask
02-924-985 125 mL Shaker Flask
02-924-988 1000 mL Shaker Flask
02-924-987 500 mL Shaker Flask
02-924-986 250 mL Shaker Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-924-989 Supplier DWK Life Sciences Supplier No. WPSFC2000
Add to Cart
Add to Cart
WHEATON™ Polypropylene Clamps for Erlenmeyer Shake Flasks are carefully engineered to provide clipping elasticity and motion resistance. Non-scratch design, standard blue color with stainless-steel screws and washers. Fit all industry standard shaking incubators. Autoclavable for reuse.
  • Carefully engineered to provide both clipping elasticity and motion resistance
  • Feature a non-scratch design
  • Offered in a standard blue color accompanied by stainless-steel screws and washers
  • Fit all industry standard shaking incubators
  • Autoclavable for reuse
TRUSTED_SUSTAINABILITY
Specifications
Holds 2000 mL Shaker Flask
Product Type Plastic Shaker Flask Clamp
Material Polypropylene
Color Blue
Quantity 1 Ea.