Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Polypropylene Pour Ring for Lab 45 Graduated Bottles
GSA_VASDP

Catalog No. 0645084 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
45 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
06-450-84 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-450-84 Supplier DWK Life Sciences Supplier No. 240756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The WHEATON™ Polypropylene Pour Ring for Lab 45 Graduated Bottles is a replaceable, clear drip-free pour ring designed for GL 45 screw thread lab bottles. It supports clean, controlled pouring and is compatible with autoclavable bottle systems.

  • Replaceable clear (natural) polypropylene pour ring for GL 45 lab bottles
  • Drip-free design supports clean, controlled pouring
  • Compatible with GL 45 screw thread graduated laboratory bottles
  • Autoclavable for use in sterilisation workflows
  • Maximum temperature: 140°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Material Polypropylene
Sterility Non-sterile
Neck Finish 45mm
For Use With (Equipment) For Lab 45's graduated bottle
Closure Size 45 mm
Autoclavable Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Seal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.