Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton PP autosampler via
Catalog No. 502905312
Encompass
0.1 mL vol. 12 x 32 mm. Lightweight nonbreakable high chemical resistance. Compatible with Beckman(R) Agilent(R) PerkinElmer(R) Spectra-Physics(R) autosamplers.1PAK 1 000 vials0.1PAK 100 vials
Catalog No. 50-290-5312 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1625151PAK
0.1 mL vol. 12 x 32 mm. Lightweight nonbreakable high chemical resistance. Compatible with Beckman(R) Agilent(R) PerkinElmer(R) Spectra-Physics(R) autosamplers.1PAK 1 000 vials0.1PAK 100 vials