Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Round Translucent LDPE Bottles
SDP

Catalog No. 0343970A Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
7 mL
15 mL
30 mL
Neck Finish:
15-415
20-410
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Neck Finish
03-439-70A 7 mL 15-415
03-439-70B 15 mL 15-415
03-439-70C 30 mL 20-410
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-439-70A Supplier DWK Life Sciences Supplier No. 221140
Add to Cart
Add to Cart

Very flexible, high impact strength plastic

Narrow mouth is ideal for liquids.

  • Excellent for mild and strong buffers, good chemical resistance
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.24 oz., 7 mL
Capacity (Metric) 7 mL
Capacity (English) 0.24 oz.
Color Translucent
Diameter (English) 0.74 in.
Material LDPE
Closure Color White
Neck Finish 15-415
Certifications/Compliance ISO 9001
Closure Included Yes
Height (Metric) 63 mm
Height (English) 2.4 in.
Closure Liner Polyethylene Foam
Closure Material Polyethylene
Closure Size 15-415 mm
Diameter (Metric) 19 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.