Learn More
Description
- Very flexible low density polyethylene (LDPE) construction with high impact strength
- Translucent, natural milky color for easy content visibility
- Narrow mouth design ideal for liquids
- Excellent chemical resistance for mild and strong buffers
- Pre-attached white polypropylene screw cap with foamed polyethylene liner
Specifications
Specifications
| Capacity | 1 oz., 30mL |
| Capacity (Metric) | 30 mL |
| Color | Translucent |
| Diameter (English) | 1.29 in. |
| Material | LDPE |
| Sterility | Non-sterile |
| Closure Color | White |
| Neck Finish | 20-410 |
| Closure Included | Yes |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
