Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton sample vials clear
Catalog No. 502900461
Encompass
Vials are packaged in corrugated trays with partitions then shrink-wrapped to reduce particulate contamination.Autoclavable borosilicate USP Type 1 glass. Order caps separately.
Catalog No. 50-290-0461 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1887001PAK
Vials are packaged in corrugated trays with partitions then shrink-wrapped to reduce particulate contamination.Autoclavable borosilicate USP Type 1 glass. Order caps separately.