Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton sample vials with
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2561961PAK
Encompass
Caps have white styrene-butadiene rubber liner and are packed in Lab-file(R) cases. Clear Wheaton-33 low extractable borosilicate glass or amber Wheaton-320 glass. Can be sterilized and stored at low temperatures.
Catalog No. 50-290-4977