Learn More
DWK Life Sciences WHEATON™ 12/2 Stainless Steel Ball Joint Clamps
Shop All DWK Life Sciences Products

Description
- Stainless steel construction for durability and corrosion resistance
- Size 12/2 ball joint clamps for compatible glassware assemblies
- Designed to securely hold ball joint glassware connections in place
- Suitable for use in standard laboratory glassware setups
Specifications
Specifications
| Quantity | 1 Cs. |
| Type | Ball Joint Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | Arsine generator |
| Color | Silver |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.