Learn More
DWK Life Sciences Wheaton™ Arsine Generator, Modified
SDP
Catalog No. 014051 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
01-405-1 24/40
1 options
Catalog No. 01-405-1 Supplier DWK Life Sciences Supplier No. 180023
Add to Cart
Add to Cart
Modified for Arsine analysis
  • Modified version of catalog number 180020 for arsine analysis
  • The freestanding system has a standard taper 24/40 Clear-Seal™ joint
  • One piece scrubber/absorber unit
  • Low extractable borosilicate glass
  • Eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes
  • Manufactured from WHEATON 33 low-extractable borosilicate glass
Compliance
Conforms to USP Type I and ASTM E 438 Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Arsine Generator
Standard Taper Joint 24/40
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For arsine analysis using SDDC method
Includes 125mL erlenmeyer flask with 24/40 joint, one-piece scrubber/absorber unit