Learn More
DWK Life Sciences Wheaton™ Arsine Generator With Clear–Seal Joint And 24/40 Standard Taper
SDP
Catalog No. 02912559 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
02-912-559 24/40
1 options
Catalog No. 02-912-559 Supplier DWK Life Sciences Supplier No. 180020
Add to Cart
Add to Cart
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
  • Manufactured from WHEATON 33 low extractable borosilicate glass
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements; Reference: £Standard Test Method for Arsenic in Water,£ASTM D2972
TRUSTED_SUSTAINABILITY
Specifications
Product Type Arsine Generator
Standard Taper Joint 24/40
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For arsine analysis using SDDC method
Includes 125mL erlenmeyer flask scrubber tube, socket joint, size 12 stainless steel ball joint clamp