Learn More
DWK Life Sciences Wheaton™ Arsine Scrubber Tube With 24/40, 12/2 Joint
SDP
Catalog No. 02912561 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40, 12/2
Catalog No. Standard Taper Joint
02-912-561 24/40, 12/2
1 options
Catalog No. 02-912-561 Supplier DWK Life Sciences Supplier No. 180022
Add to Cart
Add to Cart
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
  • Scrubber tube
  • Standard taper 24/40
  • Clear-Seal joint and a spherical 12/2 ball joint held together by a size 12 stainless steel ball joint clamp
  • Manufactured from WHEATON 33 low extractable borosilicate glass
Compliance
Meets ASTM / USP and EPA specifications; Borosilicate glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Arsine Scrubber Tube
Standard Taper Joint 24/40, 12/2
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For arsine analysis using SDDC method
For Use With (Equipment) For arsine generator
Includes Size 12 stainless steel ball joint clamp