Learn More
DWK Life Sciences WHEATON™ Erlenmeyer Flask With Screw Threads and Cap
SDP
Catalog No. 02911605 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
Catalog No. Capacity (Metric)
02-911-605 1000 mL
1 options
Catalog No. 02-911-605 Supplier DWK Life Sciences Supplier No. 635165
Add to Cart
Add to Cart
WHEATON™ Erlenmeyer Flask With Screw Threads and Cap is made from WHEATON™ 33 low extractable borosilicate glass and features an ultrasonically bonded silicone liner cap with a 38-430 screw cap size. Ideal for secure sample storage and lab use.
  • Made from Wheaton 33 low extractable borosilicate glass
  • Ultrasonically bonded silicone liner cap for a reliable, leak-resistant seal
  • 38-430 screw cap size
  • 100 mL capacity
TRUSTED_SUSTAINABILITY
Specifications
Description 1 per case
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Includes Screw threads and caps, ultrasonically bonded Silicone liner cap
Product Type Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Mouth Style Threaded
Sterility Non-sterile
Bottom Shape Flat
Surface Treatment Non-treated