Learn More
DWK Life Sciences WHEATON™ Reaction Flask With 24/40 Joint
SDP
Catalog No. 02912557 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
200 mL
Catalog No. Capacity (Metric)
02-912-557 200 mL
1 options
Catalog No. 02-912-557 Supplier DWK Life Sciences Supplier No. 635965
Add to Cart
Add to Cart
WHEATON™ Reaction Flask With 24/40 Joint is a freestanding 200mL borosilicate glass flask featuring a 24/40 standard taper joint, ideal for reaction and recovery applications requiring a secure, leak-resistant connection.
  • Freestanding design for stable bench use
  • Constructed from Wheaton 33 low extractable borosilicate glass
  • 200mL capacity
  • 24/40 standard taper joint for secure connections
  • Suitable for reaction and recovery applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 200 mL
Material Borosilicate Glass
Product Type Reaction flask
Standard Taper Joint 24/40
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Flat