Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps With PTFE Faced Foamed Polyethylene Liner
Shop All DWK Life Sciences Products

Description
- Can withstand autoclaving / sterilization
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 144 Cs. |
| Liner Material | PTFE/Polyethylene |
| Neck Finish | 15-425 |
| For Use With (Equipment) | Glass bottle or vial |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.