Description
- Designed for arsenic analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
- Manufactured from WHEATON 33 low-extractable borosilicate glass
- Assembled apparatus is freestanding and easy to use
- Design of catalog No. 02912555 eliminates the ball and socket joint connecting the scrubber and absorber components
Compliance
Conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Specifications
| Product Type | Arsine Generator |
| Material | Borosilicate Glass |
| Certifications/Compliance | USP Type I and ASTM E438 Type I, Class A |
| For Use With (Application) | For arsine analysis using SDDC method |