Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
SDP

Catalog No. p-4677519 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
PTFE/Butyl
PTFE/Silicone
Color (Seal):
Natural
Red/White
White/Transparent Blue
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum Color (Seal)
03-410-732 PTFE/Silicone White/Transparent Blue
03-410-733 PTFE/Butyl Natural
03-410-734 PTFE/Silicone Red/White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-410-732 Supplier DWK Life Sciences Supplier No. W224218
Only null left
Add to Cart
Add to Cart

Screw thread finish eliminates the need for crimping tools

  • For use with CTC Analytics Systems requiring magnetic closures
  • Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
TRUSTED_SUSTAINABILITY

Specifications

Color (Seal) White/Transparent Blue
Material PTFE, Silicone
Septum PTFE/Silicone
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.