Learn More
DWK Life Sciences Wheaton™ Septa for Open-Top Caps
SDP
Catalog No. 02927024 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
PTFE-faced Natural Rubber
PTFE-faced Red Rubber
PTFE-faced Silicone
Diameter (Metric):
8 mm
15 mm
22 mm
33 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Diameter (Metric)
02-927-024 PTFE-faced Red Rubber 8 mm
06-450-71 PTFE-faced Natural Rubber 15 mm
06-450-69 PTFE-faced Silicone 33 mm
06-450-67 PTFE-faced Silicone 22 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-927-024 Supplier DWK Life Sciences Supplier No. W240596
Add to Cart
Add to Cart
Can be used with screw caps
  • For caps with open tops
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE-faced Red Rubber
Diameter (Metric) 8 mm