Description
- Designed for use with open-top screw caps
- Autoclavable for repeated use
- Available in multiple sizes to fit a range of cap diameters
- Available in a variety of materials including PTFE-faced silicone, PTFE-faced natural rubber, and PTFE-faced natural red rubber
Specifications
Specifications
| Color | Natural |
| Material | PTFE-faced Silicone |
| Sterility | Non-sterile |
| Closure Size | 33 mm |
| Diameter (Metric) | 33 mm |
| Autoclavable | Autoclavable |
| Shape | Round |
| Product Type | Septa |