Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ Wheaton™ Serum Vial
SDP

Catalog No. 01257627 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
2 mL
3 mL
4 mL
5 mL
10 mL
Color:
Amber
Clear
Quantity:
10 Vials/Pk., 100 Pk./Cs.
100 Vials/Pk.
100 Vials/Pk., 10 Pk./Cs.
12 Vials/Pk., 196 Pk./Cs.
150 Vials/Pk.
150 Vials/Pk., 14 Pk./Cs.
169 Vials/Pk.
169 Vials/Pk., 10 Pk./Cs.
169 Vials/Pk., 304 Pk./Cs.
225 Vials/Pk.
225 Vials/Pk., 12 Pk./Cs.
225 Vials/Pk., 304 Pk./Cs.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Quantity
01-257-627 10 mL Clear 10 Vials/Pk., 100 Pk./Cs.
01-671-210 2 mL Amber 225 Vials/Pk.
01-671-211 2 mL Clear 225 Vials/Pk.
01-257-617 2 mL Clear 225 Vials/Pk., 12 Pk./Cs.
01-257-616 2 mL Clear 225 Vials/Pk., 304 Pk./Cs.
01-257-619 2 mL Amber 225 Vials/Pk., 12 Pk./Cs.
01-671-205 3 mL Clear 169 Vials/Pk.
01-257-613 3 mL Clear 169 Vials/Pk., 10 Pk./Cs.
01-671-204 3 mL Amber 169 Vials/Pk.
01-257-614 3 mL Amber 169 Vials/Pk., 304 Pk./Cs.
01-257-629 4 mL Clear 12 Vials/Pk., 196 Pk./Cs.
01-671-208 5 mL Clear 100 Vials/Pk.
01-257-609 5 mL Clear 100 Vials/Pk., 10 Pk./Cs.
01-671-206 5 mL Amber 100 Vials/Pk.
01-257-611 5 mL Amber 100 Vials/Pk., 10 Pk./Cs.
01-671-209 10 mL Clear 150 Vials/Pk.
01-257-621 10 mL Clear 150 Vials/Pk., 14 Pk./Cs.
01-671-207 10 mL Amber 150 Vials/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 01-257-627 Supplier DWK Life Sciences™ Supplier No. W0EBB06RCEP
Only null left
Add to Cart
Add to Cart

Serum vials are converted from Type I borosilicate glass tubing.

  • Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Diameter (Metric) Outer 20 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Volume (Metric) 10 mL
Closure Type Accepts 20mm aluminum seals and 13x20 stoppers (Lyo or Snap-on styles)
Quantity 10 Vials/Pk., 100 Pk./Cs.
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.