Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Sleeve Style Stoppers for Vials without Crimp Closure
SDP

Catalog No. 02985136 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-985-136 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-985-136 Supplier DWK Life Sciences Supplier No. 224100160
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Sleeve Style Stoppers for Vials without Crimp Closure feature a fold-over skirt for sealing in serum finish vials without a crimp closure, providing a reliable injection site for multiple dose entry.

  • Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure
  • Made from natural red rubber with resealable properties
  • Provides injection site for multiple dose entry
  • Durometer: 40
  • Manufactured from high-quality raw materials for exceptional dimensional stability
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Bottom 13 mm
Diameter (Metric) Outer 20 mm
Diameter (Metric) Top 20 mm
Shape Cylindrical
Sterility Non-sterile
Autoclavable Autoclavable
Product Type Sleeve Stopper
Hardness 40 Durometer
Color Red
Quantity 1000/Cs.
For Use With (Equipment) Fits 1cm I.D., 2cm O.D. mouth of vials without crimp closure
Material Rubber
Diameter (Metric) Inner 2cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.